Thermo Scientific Chemicals

1,2-Cyclohexanediol, 98%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 111130250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

1,2-Cyclohexanediol, 98%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 111130250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 111130250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
931-17-9
IUPAC Name
cyclohexane-1,2-diol
Molecular Formula
C6H12O2
InChI Key
PFURGBBHAOXLIO-UHFFFAOYNA-N
SMILES
OC1CCCCC1O
Melting point
73°C to 77°C
Appearance (Form)
Crystalline powder
GC
>=97.5 %
Appearance (Color)
White to off-white
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text