1,2-Cyclohexanediol, 98%, mixture of cis and trans, Thermo Scientific Chemicals
1,2-Cyclohexanediol, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Cyclohexanediol, 98%, mixture of cis and trans, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 111131000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS931-17-9
IUPAC Namecyclohexane-1,2-diol
Molecular FormulaC6H12O2
InChI KeyPFURGBBHAOXLIO-UHFFFAOYNA-N
SMILESOC1CCCCC1O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point73°C to 77°C
Appearance (Form)Crystalline powder
GC>=97.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only