1,4-Cyclohexanediol, 99%, mixture of cis and trans, Thermo Scientific Chemicals
1,4-Cyclohexanediol, 99%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Cyclohexanediol, 99%, mixture of cis and trans, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 111151000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS556-48-9
IUPAC Namecyclohexane-1,4-diol
Molecular FormulaC6H12O2
InChI KeyVKONPUDBRVKQLM-UHFFFAOYSA-N
SMILESOC1CCC(O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point97°C to 102°C
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only