Thermo Scientific Chemicals

1,2-Cyclohexanedione, 98%, Thermo Scientific Chemicals

Catalog number: 111160500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

1,2-Cyclohexanedione, 98%, Thermo Scientific Chemicals

Catalog number: 111160500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 111160500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
765-87-7
IUPAC Name
cyclohexane-1,2-dione
Molecular Formula
C6H8O2
InChI Key
OILAIQUEIWYQPH-UHFFFAOYSA-N
SMILES
O=C1CCCCC1=O
Appearance (Color)
Clear yellow
Infrared spectrum
Conforms
Refractive index
1.5180 to 1.5200 (20°C, 589 nm)
Appearance (Form)
Liquid after melting
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text