1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 111171000
Price (USD)/ Each
54.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
64.50 
Save 9.85 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
54.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
64.50 
Save 9.85 (15%)
Add to cart
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
Catalog number111171000
Price (USD)/ Each
54.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
64.50 
Save 9.85 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namecyclohexane-1,3-dione
Molecular FormulaC6H8O2
InChI KeyHJSLFCCWAKVHIW-UHFFFAOYSA-N
SMILESO=C1CCCC(=O)C1
Molecular Weight (g/mol)112.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Water=<0.5 %
Appearance (Color)Light yellow to beige
Melting point>=101°C
Titration Argentometric=<1 % Sodium chloride
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only