Cyclohexanone, 99.8%, extra pure
Cyclohexanone, 99.8%, extra pure
Cyclohexanone, 99.8%, extra pure
Cyclohexanone, 99.8%, extra pure
Thermo Scientific Chemicals

Cyclohexanone, 99.8%, extra pure

CAS: 108-94-1 | C6H10O | 98.145 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1111900101 LGlass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 111190010
Price (USD)
49.65
Special offer
Online exclusive
Ends: 26-Sep-2025
57.90
Save 8.25 (14%)
Each
In stock
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 26-Sep-2025
57.90
Save 8.25 (14%)
Each
Add to cart
Chemical Identifiers
CAS108-94-1
IUPAC Namecyclohexanone
Molecular FormulaC6H10O
InChI KeyJHIVVAPYMSGYDF-UHFFFAOYSA-N
SMILESO=C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<20 APHA
Infrared spectrumConforms
GC>=99.75 %
Water=<0.2 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link