Thermo Scientific Chemicals

Cyclohexylacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 111270050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Cyclohexylacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 111270050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 111270050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5292-21-7
IUPAC Name
2-cyclohexylacetic acid
Molecular Formula
C8H14O2
InChI Key
LJOODBDWMQKMFB-UHFFFAOYSA-N
SMILES
OC(=O)CC1CCCCC1
Appearance (Color)
White to light yellow
Melting point
27°C to 31°C
Infrared spectrum
Conforms
Titration with NaOH
>=98.0 %
Appearance (Form)
Low melting solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text