Thermo Scientific Chemicals

1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimide metho-p-toluenesulfonate, 95%, Thermo Scientific Chemicals

Catalog number: 111360250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimide metho-p-toluenesulfonate, 95%, Thermo Scientific Chemicals

Catalog number: 111360250
25 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 111360250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2491-17-0
IUPAC Name
4-(2-{[(cyclohexylimino)methylidene]amino}ethyl)-4-methylmorpholin-4-ium 4-methylbenzene-1-sulfonate
Molecular Formula
C21H33N3O4S
InChI Key
GBCAVSYHPPARHX-UHFFFAOYSA-M
SMILES
CC1=CC=C(C=C1)S([O-])(=O)=O.C[N+]1(CCN=C=NC2CCCCC2)CCOCC1
Appearance (Color)
White to light yellow
Appearance (Form)
Powder and/or chunks
Melting point
112°C to 120°C
HPLC
>=94 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text