Nutritional Supplements
<p>Browse through our collection of nutritional supplements designed to enhance cell culture media. These supplements provide essential nourishment and support for cell growth in various applications. Our range includes amino acid supplements, bacterial media, casamino acids, dehydrated culture media, glutamine products, yeast extracts, peptones, and other specialized supplements. These products are available in different forms such as powders, liquids, and dehydrated granules, catering to diverse research and production needs.Nutritional supplements for cell culture media encompass a wide array of components, from basic amino acids to complex yeast-derived extracts. They are crucial for optimizing cell growth, protein production, and overall culture performance. Many of these supplements are animal origin-free, chemically defined, or protein-free, meeting the stringent requirements of modern biotechnology and pharmaceutical applications.Examples of products in this category include tryptone, yeast extracts, proteose peptones, GlutaMAX supplement, casitone, and malt extract. These supplements can be used in mammalian cell culture, microbial fermentation, and other biotechnological processes to enhance cell growth, productivity, and product quality. Learn more about <u><b><a href='usenhomelife-sciencecell-culturemammalian-cell-culturemedia-supplements.html#more-media-supplements'>cell culture media supplements<a><b><u>.<p>