Thermo Scientific Chemicals

Cyclopentane, 97%, extra pure, Thermo Scientific Chemicals

Catalog number: 111480025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Cyclopentane, 97%, extra pure, Thermo Scientific Chemicals

Catalog number: 111480025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 111480025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
287-92-3
IUPAC Name
cyclopentane
Molecular Formula
C5H10
InChI Key
RGSFGYAAUTVSQA-UHFFFAOYSA-N
SMILES
C1CCCC1
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
GC
>=96.0 %
Residue after evaporation
=<0.01 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text