Ciclopentano, 97 %, extra puro, Thermo Scientific Chemicals
Ciclopentano, 97 %, extra puro, Thermo Scientific Chemicals
Ciclopentano, 97 %, extra puro, Thermo Scientific Chemicals
Ciclopentano, 97 %, extra puro, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ciclopentano, 97 %, extra puro, Thermo Scientific Chemicals

Ciclopentano, 97 %, C5H10, número CAS-287-92-3, ciclopentano, ciclopentano, ciclopentanos, grupo ciclopentilo, hsdb 62, hidrocarburos, c5 y c6 cíclicos, pentametileno, t86pb90rnu, unii-t86pb90rnu, ciclopentano, 1 l, 96 % mín. (GC), 05, 19, 50,0 °C, CHEBI:23492, blanco, 0,7500 g/ml, 206-016-6, -7 °C
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 L
100 mL
2.5 L
Envase:
Frasco de vidrio
Número de catálogo 111481000
Precio (EUR)
34,43
Precio exclusivo en nuestra web
40,50
Ahorro 6,07 (15%)
Each
-
Añadir al carro de la compra
Cantidad:
100 mL
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
34,43
Precio exclusivo en nuestra web
40,50
Ahorro 6,07 (15%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS287-92-3
IUPAC Namecyclopentane
Molecular FormulaC5H10
InChI KeyRGSFGYAAUTVSQA-UHFFFAOYSA-N
SMILESC1CCCC1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
GC>=96.0 %
Residue after evaporation=<0.01 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
Ver más
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Los clientes que vieron este artículo también vieron



Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
A0467155Certificate of Analysis25 oct 2024111480025, 111481000, 111480010
A0455415Certificate of Analysis25 ago 2023111480025, 111481000, 111480010
A0408397Certificate of Analysis13 dic 2022111480025, 111481000, 111480010
A0446902Certificate of Analysis24 oct 2022111480025, 111481000, 111480010
A0426164Certificate of Analysis14 ene 2022111480025, 111481000, 111480010
Se muestran 5 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable liquids
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Aspiration Toxicity
Category 1
Chronic aquatic toxicity
Category 3
Label Elements
Signal Word

Danger

Hazard Statements

H304 - May be fatal if swallowed and enters airways

H336 - May cause drowsiness or dizziness

H412 - Harmful to aquatic life with long lasting effects

Physical Hazards

H225 - Highly flammable liquid and vapor

EU Specific Hazard Statements

EUH066 - Repeated exposure may cause skin dryness or cracking

Precautionary Statements

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P331 - Do NOT induce vomiting

Compartir número de catálogo, nombre o enlace.

1x1 image pixel for data collection