Cyclopentanecarboxylic Acid, 98+%, Thermo Scientific Chemicals
Cyclopentanecarboxylic Acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopentanecarboxylic Acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 111491000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3400-45-1
IUPAC Namecyclopentanecarboxylic acid
Molecular FormulaC6H10O2
InChI KeyJBDSSBMEKXHSJF-UHFFFAOYSA-N
SMILESOC(=O)C1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with NaOH98.0 to 102.0 %
GC>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only