Cyclopentanol, 99%, Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopentanol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 111525000
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS96-41-3
IUPAC Namecyclopentanol
Molecular FormulaC5H10O
InChI KeyXCIXKGXIYUWCLL-UHFFFAOYSA-N
SMILESOC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4520 to 1.4540 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only