Cyclopentanone, 99+%, pure, Thermo Scientific Chemicals
Cyclopentanone, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopentanone, 99+%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
10 L
Packaging:
View all
Glass bottle
Catalog number 111530025
Price (USD)/ Each
467.65
Online exclusive
519.00 
Save 51.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
467.65
Online exclusive
519.00 
Save 51.35 (10%)
Add to cart
Cyclopentanone, 99+%, pure, Thermo Scientific Chemicals
Catalog number111530025
Price (USD)/ Each
467.65
Online exclusive
519.00 
Save 51.35 (10%)
-
Add to cart
Chemical Identifiers
CAS120-92-3
IUPAC Namecyclopentanone
Molecular FormulaC5H8O
InChI KeyBGTOWKSIORTVQH-UHFFFAOYSA-N
SMILESO=C1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Refractive index1.4360 to 1.4380 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only