Thermo Scientific Chemicals

Cyclopentanone, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 111530100
10 L, Plastic drum, Each
Thermo Scientific Chemicals

Cyclopentanone, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 111530100
10 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 111530100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
120-92-3
IUPAC Name
cyclopentanone
Molecular Formula
C5H8O
InChI Key
BGTOWKSIORTVQH-UHFFFAOYSA-N
SMILES
O=C1CCCC1
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=99.0 %
Refractive index
1.4360 to 1.4380 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text