Invitrogen™

Dynabeads™ CD31 Endothelial Cell

Catalog number: 11155D
Have Questions?
Invitrogen™

Dynabeads™ CD31 Endothelial Cell

Catalog number: 11155D
Have Questions?
Catalog Number
11155D
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
11155D5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Isolate endothelial cells directly from tissue digests. Dynabeads™ CD31 are ideal for positively isolating endothelial cells for downstream molecular analysis.

• Positively isolate endothelial cells directly from tissue digests for molecular applications
• Establish cultures for endothelial cells
• No columns required

Starting sample:
Tissue digests.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Endothelial Cells
Product Type
Antibody Coated Bead
Isolation Technology
Positive Isolation
No. of Cells
Processes ∼2 x 109 cells total
Output Viability
>95%
Product Line
DYNAL™, Dynabeads™
Purity or Quality Grade
Research Grade
Sample Type
Tissue Digests
Shipping Condition
Room Temperature
Starting Material Cell No.
1 x 108 cells per isolation
Target Species
Human

Contents & Storage

This Product Contains: 5 mL Dynabeads™ coated with anti-CD31 monoclonal antibody.

Store at 2°C to 8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text