Thermo Scientific Chemicals

Cyclopentylamine, 99+%, Thermo Scientific Chemicals

Catalog number: 111590250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Cyclopentylamine, 99+%, Thermo Scientific Chemicals

Catalog number: 111590250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 111590250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1003-03-8
IUPAC Name
cyclopentanamine
Molecular Formula
C5H11N
InChI Key
NISGSNTVMOOSJQ-UHFFFAOYSA-N
SMILES
NC1CCCC1
Color scale
=<50 APHA
Infrared spectrum
Conforms
Water
=<0.3 %
Appearance (Form)
Clear liquid
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text