Thermo Scientific Chemicals

Cyclopentylamine, 99+%, Thermo Scientific Chemicals

Catalog number: 111591000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Cyclopentylamine, 99+%, Thermo Scientific Chemicals

Catalog number: 111591000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 111591000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1003-03-8
IUPAC Name
cyclopentanamine
Molecular Formula
C5H11N
InChI Key
NISGSNTVMOOSJQ-UHFFFAOYSA-N
SMILES
NC1CCCC1
Color scale
=<50 APHA
Appearance (Form)
Clear liquid
GC
>=99.0 %
Refractive index
1.4482 to 1.4502 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text