Cyclopentyl bromide, 98%, Thermo Scientific Chemicals
Cyclopentyl bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopentyl bromide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 111600010
Price (USD)/ Each
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namebromocyclopentane
Molecular FormulaC5H9Br
InChI KeyBRTFVKHPEHKBQF-UHFFFAOYSA-N
SMILESBrC1CCCC1
Molecular Weight (g/mol)149.03
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4870 to 1.4910 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only