Thermo Scientific Chemicals

Cyclopropanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals

Catalog number: 111631000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Cyclopropanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals

Catalog number: 111631000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 111631000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4023-34-1
IUPAC Name
cyclopropanecarbonyl chloride
Molecular Formula
C4H5ClO
InChI Key
ZOOSILUVXHVRJE-UHFFFAOYSA-N
SMILES
ClC(=O)C1CC1
Appearance (Color)
Clear colorless to light yellow
GC
>=97.5 %
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Titration RCOCl
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text