Thermo Scientific Chemicals

Cyclopropyl methyl ketone, 98%, Thermo Scientific Chemicals

Catalog number: 111715000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

Cyclopropyl methyl ketone, 98%, Thermo Scientific Chemicals

Catalog number: 111715000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 111715000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
765-43-5
IUPAC Name
1-cyclopropylethan-1-one
Molecular Formula
C5H8O
InChI Key
HVCFCNAITDHQFX-UHFFFAOYSA-N
SMILES
CC(=O)C1CC1
Infrared spectrum
Conforms
Water
=<0.2 %
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text