Thermo Scientific Chemicals

Cystamine Dihydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 111771000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Cystamine Dihydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 111771000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 111771000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-17-7
IUPAC Name
dihydrogen 2-[(2-aminoethyl)disulfanyl]ethan-1-amine dichloride
Molecular Formula
C4H14Cl2N2S2
InChI Key
YUFRRMZSSPQMOS-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NCCSSCCN
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals and/or chunks
Titration Argentometric
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text