Thermo Scientific Chemicals

Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 111840025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 111840025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 111840025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
91-17-8
IUPAC Name
decahydronaphthalene
Molecular Formula
C10H18
InChI Key
NNBZCPXTIHJBJL-UHFFFAOYSA-N
SMILES
C1CCC2CCCCC2C1
Color scale
=<10 APHA
Refractive index
1.4730 to 1.4760 (20°C, 589 nm)
Infrared spectrum
Conforms
Water
=<0.1 %
Appearance (Form)
Clear liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text