Decane, 99+%, pure, Thermo Scientific Chemicals
Decane, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Decane, 99+%, pure, Thermo Scientific Chemicals

Decane, >-99.0%, C10H22, CAS Number-124-18-5, ccris 653, decane, analytical standard, decyl hydride, dsstox_cid_4913, dsstox_gsid_24913, dsstox_rid_77577, hsdb 63, n-decane, nonane, methyl, unii-nk85062oiy, 2.5L, 99+%, 01, 168, CHEBI:41808, Colorless, 0.7350g/mL, 204-686-4, 142.28, Pure, Danger
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 111871000
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.50 
Save 8.85 (14%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.50 
Save 8.85 (14%)
Add to cart
Decane, 99+%, pure, Thermo Scientific Chemicals
Catalog number111871000
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.50 
Save 8.85 (14%)
-
Add to cart
Chemical Identifiers
CAS124-18-5
IUPAC Namedecane
Molecular FormulaC10H22
InChI KeyDIOQZVSQGTUSAI-UHFFFAOYSA-N
SMILESCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.0 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4100 to 1.4130 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only