Thermo Scientific Chemicals

1,10-Decanedicarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: 111885000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

1,10-Decanedicarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: 111885000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 111885000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
693-23-2
IUPAC Name
dodecanedioic acid
Molecular Formula
C12H22O4
InChI Key
TVIDDXQYHWJXFK-UHFFFAOYSA-N
SMILES
OC(=O)CCCCCCCCCCC(O)=O
Appearance (Form)
Powder or pellets
Titration with NaOH
>=98.5 %
Appearance (Color)
White
Infrared spectrum
Conforms
Melting point
125°C to 130°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text