Thermo Scientific Chemicals

Dehydroacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 111941000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Dehydroacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 111941000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 111941000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
520-45-6
IUPAC Name
3-acetyl-6-methyl-3,4-dihydro-2H-pyran-2,4-dione
Molecular Formula
C8H8O4
InChI Key
PGRHXDWITVMQBC-UHFFFAOYNA-N
SMILES
CC(=O)C1C(=O)OC(C)=CC1=O
Appearance (Color)
Cream to light yellow
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %
Melting point
109°C to 112°C
Sulfated ash
=<0.1 % (700°C, 2 h)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text