Thermo Scientific Chemicals

Deoxybenzoin, 98%, Thermo Scientific Chemicals

Catalog number: 111961000
100 g, Each
Thermo Scientific Chemicals

Deoxybenzoin, 98%, Thermo Scientific Chemicals

Catalog number: 111961000
100 g, Each
Quantity
Catalog number: 111961000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
451-40-1
IUPAC Name
1,2-diphenylethan-1-one
Molecular Formula
C14H12O
InChI Key
OTKCEEWUXHVZQI-UHFFFAOYSA-N
SMILES
O=C(CC1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Powder or crystalline powder or crystals
Appearance (Color)
White to yellow to light brown
Appearance (Form)
and/or chunks
Melting point
51°C to 58°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text