Thermo Scientific Chemicals

Diacetone-D-glucose, 98+%

Catalog number: 112011000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Diacetone-D-glucose, 98+%

Catalog number: 112011000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 112011000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
582-52-5
IUPAC Name
5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol
Molecular Formula
C12H20O6
InChI Key
KEJGAYKWRDILTF-UHFFFAOYNA-N
SMILES
CC1(C)OCC(O1)C1OC2OC(C)(C)OC2C1O
Appearance (Form)
Powder
GC
>=98.0 %
Sulfated ash
=<0.2 %
Appearance (Color)
White to yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text