Diallylamine, 97%, Thermo Scientific Chemicals
Diallylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diallylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Catalog number 112040010
Price (USD)/ Each
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS124-02-7
IUPAC Namebis(prop-2-en-1-yl)amine
Molecular FormulaC6H11N
InChI KeyDYUWTXWIYMHBQS-UHFFFAOYSA-N
SMILESC=CCNCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.4395 to 1.4415 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only