Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Search Thermo Fisher Scientific
Catalog Number | Type |
---|---|
11206175 | CN IRMS System |
11206180 | CN/OH IRMS System |
11206185 | CNSOH IRMS System |
BRE723644 | OH IRMS System |
Discover an automated, easy-to-use solution for μg- to mg-level elemental and isotope analysis with the Thermo Scientific™ EA IsoLink™ IRMS System. This all-in-one isotope ratio mass spectrometry system enables laboratories to increase sample throughput, reduce cost per sample, and provides high-precision elemental and isotopic analysis. Innovation in the EA IsoLink IRMS System provides helium saving technology and integration of temperature ramped gas chromatography delivering quick analysis times and low cost analysis alongside outstanding quality data, especially on small sample amounts. These features allow you to push boundaries for research and routine analyses in ecology, biology, archaeology, geosciences, food integrity, and forensics whilst offering built-in flexibility to quickly and simply adapt to changing analytical demands.
The EA IsoLink IRMS System uses temperature-ramped gas chromatography (GC), which reduces analysis time per sample, increases sample throughput, and produces high quality data thanks to optimal baseline separation of analyte gases and sharp peak shapes. These benefits are the prerequisites for high-performance IRMS.
Low Helium ConsumptionUse the Thermo Scientific™ Intelligent Helium Management (HeM) Module to ensure low helium consumption during your analysis and significantly reduce your sample analysis costs.
EA IsoLink IRMS System FeaturesTracking origins, tracing identity: the Isotope Hunter follows the clues that history leaves in foods or fibers, liquids or stone. Capture your samples’ stories with the Thermo Scientific Isotope Ratio Mass Spectrometry portfolio.
Share catalog number, name or link