EA IsoLink™ IRMS System
EA IsoLink™ IRMS System
Actual product may vary
Thermo Scientific™
EA IsoLink™ IRMS System
Get higher throughput and precision for your C, N, S, O, and H isotope ratio measurements using the Thermo Scientific™ EA IsoLink IRMS System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
11206180CN/OH IRMS System
11206175CN IRMS System
11206185CNSOH IRMS System
BRE723644OH IRMS System
Catalog number 11206180
Price (USD)
-
Request A Quote
Type:
CN/OH IRMS System
Ask our AI about this Product
Discover an automated, easy-to-use solution for μg- to mg-level elemental and isotope analysis with the Thermo Scientific™ EA IsoLink™ IRMS System. This all-in-one isotope ratio mass spectrometry system enables laboratories to increase sample throughput, reduce cost per sample, and provides high-precision elemental and isotopic analysis. Innovation in the EA IsoLink IRMS System provides helium saving technology and integration of temperature ramped gas chromatography delivering quick analysis times and low cost analysis alongside outstanding quality data, especially on small sample amounts. These features allow you to push boundaries for research and routine analyses in ecology, biology, archaeology, geosciences, food integrity, and forensics whilst offering built-in flexibility to quickly and simply adapt to changing analytical demands.
The EA IsoLink IRMS System uses temperature-ramped gas chromatography (GC), which reduces analysis time per sample, increases sample throughput, and produces high quality data thanks to optimal baseline separation of analyte gases and sharp peak shapes. These benefits are the prerequisites for high-performance IRMS.
Low Helium Consumption
Use the Thermo Scientific™ Intelligent Helium Management (HeM) Module to ensure low helium consumption during your analysis and significantly reduce your sample analysis costs.
EA IsoLink IRMS System Features
  • Sharp peak shapes and complete peak separation with temperature-ramped gas chromatography.
  • Full insight and control over the entire analytical workflow.
  • Widest dynamic range in EA-IRMS with multi-element analysis.
  • Lowest helium consumption and highest sample throughput.
  • Full system automation and lowest operating cost.
  • Flexibility for all applications with an easy-to-use modular system.
  • Unmatched robustness.
Specifications
IncludesFlash IRMS Elemental Analyzer, Delta Q IRMS, Conflo IV Universal Interface
Detector TypeThermal Conductivity Detector, IRMS
For Use With (Application)Geoscience, Food Safety, Environmental Science, Forensics, Petroleum, Exploration, Marine Science, Agronomy
TypeCN/OH IRMS System
Unit SizeEach
Investigating authenticity and tracing origin with isotope fingerprints
Investigating authenticity and tracing origin with isotope fingerprints
Tracking origins, tracing identity: the Isotope Hunter follows the clues that history leaves in foods or fibers, liquids or stone. Capture your samples’ stories with the Thermo Scientific Isotope Ratio Mass Spectrometry portfolio.
Investigate now ›
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Videos
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Scientific Resources
Product Information
Share catalog number, name or link
1x1 image pixel for data collection