Thermo Scientific Chemicals

3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 112105000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 112105000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 112105000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
619-05-6
IUPAC Name
3,4-diaminobenzoate
Molecular Formula
C7H7N2O2
InChI Key
HEMGYNNCNNODNX-UHFFFAOYSA-M
SMILES
NC1=CC=C(C=C1N)C([O-])=O
Appearance (Color)
Brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=96.0 %
Titration with NaOH
96.0 to 104.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text