Thermo Scientific Chemicals

3,5-Diaminobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 112111000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

3,5-Diaminobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 112111000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 112111000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
535-87-5
IUPAC Name
5-carboxybenzene-1,3-bis(aminium) dichloride
Molecular Formula
C7H10Cl2N2O2
InChI Key
IGPLRBRBTUNCRT-UHFFFAOYSA-N
SMILES
[Cl-].[Cl-].[NH3+]C1=CC(=CC([NH3+])=C1)C(O)=O
Appearance (Color)
White to grey
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with TBAH
97.5 to 102.5 %
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text