Thermo Scientific Chemicals

1,10-Diaminodecane, 97%, Thermo Scientific Chemicals

Catalog number: 112130250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1,10-Diaminodecane, 97%, Thermo Scientific Chemicals

Catalog number: 112130250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 112130250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
646-25-3
IUPAC Name
decane-1,10-diamine
Molecular Formula
C10H24N2
InChI Key
YQLZOAVZWJBZSY-UHFFFAOYSA-N
SMILES
NCCCCCCCCCCN
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder or flakes and/or chunks
Infrared spectrum
Conforms
Melting point
59°C to 64°C
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text