1,7-Diaminoheptane, 98%, Thermo Scientific Chemicals
1,7-Diaminoheptane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,7-Diaminoheptane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 112200050
Price (USD)/ Each
33.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
39.30 
Save 5.65 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
33.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
39.30 
Save 5.65 (14%)
Add to cart
1,7-Diaminoheptane, 98%, Thermo Scientific Chemicals
Catalog number112200050
Price (USD)/ Each
33.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
39.30 
Save 5.65 (14%)
-
Add to cart
Chemical Identifiers
CAS646-19-5
IUPAC Nameheptane-1,7-diamine
Molecular FormulaC7H18N2
InChI KeyPWSKHLMYTZNYKO-UHFFFAOYSA-N
SMILESNCCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid or solid
Infrared spectrumConforms
Titration with HClO497.5 to 102.5 %
GC>=97.5 %
Appearance (Color)Colorless to white to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only