Thermo Scientific Chemicals

1,5-Diaminonaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: 112290250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1,5-Diaminonaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: 112290250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 112290250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2243-62-1
IUPAC Name
naphthalene-1,5-diamine
Molecular Formula
C10H10N2
InChI Key
KQSABULTKYLFEV-UHFFFAOYSA-N
SMILES
NC1=CC=CC2=C(N)C=CC=C12
GC
>=96.0 %
Appearance (Color)
Grey to dark brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text