1,8-Diaminooctane, 98%, Thermo Scientific Chemicals
1,8-Diaminooctane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,8-Diaminooctane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 112310250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nameoctane-1,8-bis(aminium) dichloride
Molecular FormulaC8H22Cl2N2
InChI KeyZFLWZOGXFQNIMT-UHFFFAOYSA-N
SMILES[Cl-].[Cl-].[NH3+]CCCCCCCC[NH3+]
Molecular Weight (g/mol)217.18
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point50°C to 54°C
Titration with HClO4>=97.5 %
GC>=97.5 %
Appearance (Color)White to yellow
Appearance (Form)Fused crystalline mass
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only