1,5-Diaminopentane, 98%, Thermo Scientific Chemicals
1,5-Diaminopentane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,5-Diaminopentane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number 112320250
Price (USD)/ Each
362.65
Online exclusive
403.00 
Save 40.35 (10%)
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
362.65
Online exclusive
403.00 
Save 40.35 (10%)
Add to cart
1,5-Diaminopentane, 98%, Thermo Scientific Chemicals
Catalog number112320250
Price (USD)/ Each
362.65
Online exclusive
403.00 
Save 40.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Namepentane-1,5-diamine
Molecular FormulaC5H14N2
InChI KeyVHRGRCVQAFMJIZ-UHFFFAOYSA-N
SMILESNCCCCCN
Molecular Weight (g/mol)102.18
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Refractive index1.4570 to 1.4600 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only