1,3-Diaminopropane, 99%, Thermo Scientific Chemicals
1,3-Diaminopropane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Diaminopropane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 112350010
Price (USD)/ Each
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS109-76-2
IUPAC Namepropane-1,3-diamine
Molecular FormulaC3H10N2
InChI KeyXFNJVJPLKCPIBV-UHFFFAOYSA-N
SMILESNCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
Titration with HClO4>=98.5 %
GC>=98.5 %
Water=<0.5 %
Refractive index1.4565 to 1.4585 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only