Thermo Scientific Chemicals

2,6-Diaminopyridine, 98%, Thermo Scientific Chemicals

Catalog number: 112381000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2,6-Diaminopyridine, 98%, Thermo Scientific Chemicals

Catalog number: 112381000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 112381000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
141-86-6
IUPAC Name
pyridine-2,6-diamine
Molecular Formula
C5H7N3
InChI Key
VHNQIURBCCNWDN-UHFFFAOYSA-N
SMILES
NC1=CC=CC(N)=N1
Appearance (Color)
Beige to grey to dark brown
Appearance
May darken on storage
Infrared spectrum
Conforms
Melting point
115°C to 123°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text