Dibenzosuberenone, 97%, Thermo Scientific Chemicals
Dibenzosuberenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dibenzosuberenone, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number 112531000
Price (USD)/ Each
160.65
Online exclusive
189.00 
Save 28.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
160.65
Online exclusive
189.00 
Save 28.35 (15%)
Add to cart
Dibenzosuberenone, 97%, Thermo Scientific Chemicals
Catalog number112531000
Price (USD)/ Each
160.65
Online exclusive
189.00 
Save 28.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-one
Molecular FormulaC15H10O
InChI KeySNVTZAIYUGUKNI-UHFFFAOYSA-N
SMILESO=C1C2=CC=CC=C2C=CC2=CC=CC=C12
Molecular Weight (g/mol)206.24
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point85°C to 91°C
GC>=96.0 %
Loss on drying=<0.5 % (60°C) (vacuum)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only