Thermo Scientific Chemicals

Dibenzothiophene, 98%, Thermo Scientific Chemicals

Catalog number: 112540250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Dibenzothiophene, 98%, Thermo Scientific Chemicals

Catalog number: 112540250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 112540250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
132-65-0
IUPAC Name
8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Formula
C12H8S
InChI Key
IYYZUPMFVPLQIF-UHFFFAOYSA-N
SMILES
S1C2=C(C=CC=C2)C2=C1C=CC=C2
Appearance (Color)
White to beige to grey
Appearance (Form)
Powder or crystals and/or chunks
Infrared spectrum
Conforms
Melting point
97°C to 101°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text