1x1 image pixel for data collection
trans-1,2-Dibenzoylethylene, 97%
trans-1,2-Dibenzoylethylene, 97%
trans-1,2-Dibenzoylethylene, 97%
trans-1,2-Dibenzoylethylene, 97%
Thermo Scientific Chemicals

trans-1,2-Dibenzoylethylene, 97%

CAS: 959-28-4 | C16H12O2 | 236.27 g/mol
Have Questions?
Catalog NumberQuantity
11258025025 g
Catalog number 112580250
Price (USD)
361.00
Each
Estimated availability date 25-Jul-2025
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
361.00
Each
Add to cart
Chemical Identifiers
CAS959-28-4
IUPAC Name(2E)-1,4-diphenylbut-2-ene-1,4-dione
Molecular FormulaC16H12O2
InChI KeyWYCXGQSQHAXLPK-VAWYXSNFSA-N
SMILESO=C(\C=C\C(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to light orange
Infrared spectrumConforms
Melting point108°C to 112°C
GC>=96.0 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection