Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dibenzoylmethane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 112601000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS120-46-7
IUPAC Name1,3-diphenylpropane-1,3-dione
Molecular FormulaC15H12O2
InChI KeyNZZIMKJIVMHWJC-UHFFFAOYSA-N
SMILESO=C(CC(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Infrared spectrumConforms
Melting point75°C to 79°C
GC>=97.5 %
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only