Dibenzyl phosphate, 98%, Thermo Scientific Chemicals
Dibenzyl phosphate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dibenzyl phosphate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 112650050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1623-08-1
IUPAC Namebis(benzyloxy)phosphinic acid
Molecular FormulaC14H15O4P
InChI KeyHDFFVHSMHLDSLO-UHFFFAOYSA-N
SMILESOP(=O)(OCC1=CC=CC=C1)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point76°C to 80°C
Titration with NaOH>=97.5 %
Appearance (Form)Powder
Infrared spectrumConforms
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only