Thermo Scientific Chemicals

Dibenzyl phosphite, 95%, Thermo Scientific Chemicals

Catalog number: 112660250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Dibenzyl phosphite, 95%, Thermo Scientific Chemicals

Catalog number: 112660250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 112660250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17176-77-1
IUPAC Name
bis(benzyloxy)(oxo)-λ⁵-phosphanylium
Molecular Formula
C14H14O3P
InChI Key
RQKYHDHLEMEVDR-UHFFFAOYSA-N
SMILES
O=[P+](OCC1=CC=CC=C1)OCC1=CC=CC=C1
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.5520 to 1.5550 (20°C, 589 nm)
NMR
>=90.0 %
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text