Thermo Scientific Chemicals

1,4-Dibromobenzene, 99%, Thermo Scientific Chemicals

Catalog number: 112730025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

1,4-Dibromobenzene, 99%, Thermo Scientific Chemicals

Catalog number: 112730025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 112730025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
106-37-6
IUPAC Name
1,4-dibromobenzene
Molecular Formula
C6H4Br2
InChI Key
SWJPEBQEEAHIGZ-UHFFFAOYSA-N
SMILES
BrC1=CC=C(Br)C=C1
Appearance (Color)
Colorless to off-white
Appearance (Form)
Crystals
Infrared spectrum
Conforms
Melting point
86°C to 89°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text