1,4-Dibromobenzene, 99%, Thermo Scientific Chemicals
1,4-Dibromobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Dibromobenzene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 112732500
Price (USD)/ Each
73.65
Online exclusive
81.40 
Save 7.75 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
73.65
Online exclusive
81.40 
Save 7.75 (10%)
Add to cart
1,4-Dibromobenzene, 99%, Thermo Scientific Chemicals
Catalog number112732500
Price (USD)/ Each
73.65
Online exclusive
81.40 
Save 7.75 (10%)
-
Add to cart
Chemical Identifiers
CAS106-37-6
IUPAC Name1,4-dibromobenzene
Molecular FormulaC6H4Br2
InChI KeySWJPEBQEEAHIGZ-UHFFFAOYSA-N
SMILESBrC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to off-white
Appearance (Form)Crystals
Infrared spectrumConforms
Melting point86°C to 89°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only