Thermo Scientific Chemicals

1,10-Dibromodecane, 97%, Thermo Scientific Chemicals

Catalog number: 112781000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

1,10-Dibromodecane, 97%, Thermo Scientific Chemicals

Catalog number: 112781000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 112781000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4101-68-2
IUPAC Name
1,10-dibromodecane
Molecular Formula
C10H20Br2
InChI Key
GTQHJCOHNAFHRE-UHFFFAOYSA-N
SMILES
BrCCCCCCCCCCBr
Appearance (Color)
White to light brown
Appearance (Form)
Fused solid or liquid
Infrared spectrum
Conforms
Melting point
25°C to 30°C
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text