1,10-Dibromodecane, 97%, Thermo Scientific Chemicals
1,10-Dibromodecane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,10-Dibromodecane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 112785000
Price (USD)/ Each
505.00
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
505.00
Add to cart
1,10-Dibromodecane, 97%, Thermo Scientific Chemicals
Catalog number112785000
Price (USD)/ Each
505.00
-
Add to cart
Chemical Identifiers
IUPAC Name1,10-dibromodecane
Molecular FormulaC10H20Br2
InChI KeyGTQHJCOHNAFHRE-UHFFFAOYSA-N
SMILESBrCCCCCCCCCCBr
Molecular Weight (g/mol)300.08
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Fused solid or liquid
Infrared spectrumConforms
Melting point25°C to 30°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only